Learn More
MSC Justrite Pipe Leak Diverter, 7 in Square, PVC, 1 ea
Catalog No. 5000781582
Encompass
This leak diverter features durable, puncture-resistant, and watertight PVC coated yellow fabric, designed to channel overhead pipe and ceiling leaks to containers or floor drains. It provides an economical solution for protecting personnel, equipment, and inventory, allowing safe drainage via a standard garden hose at a flow rate of 3.3 gal/min (12.5 L/min).
  • Durable, puncture-resistant, watertight PVC fabric
  • Channels overhead pipe and ceiling leaks
  • Protects personnel, equipment, and inventory
  • Connects to standard garden hose
  • Flow rate of 3.3 gal/min (12.5 L/min)
  • Pipe leak diverter
  • PVC material
  • Square shape
  • Yellow color
  • Includes gromets
Catalog No. 50-007-81582 Supplier MSC Supplier No. 53807194
This leak diverter features durable, puncture-resistant, and watertight PVC coated yellow fabric, designed to channel overhead pipe and ceiling leaks to containers or floor drains. It provides an economical solution for protecting personnel, equipment, and inventory, allowing safe drainage via a standard garden hose at a flow rate of 3.3 gal/min (12.5 L/min).
  • Durable, puncture-resistant, watertight PVC fabric
  • Channels overhead pipe and ceiling leaks
  • Protects personnel, equipment, and inventory
  • Connects to standard garden hose
  • Flow rate of 3.3 gal/min (12.5 L/min)
  • Pipe leak diverter
  • PVC material
  • Square shape
  • Yellow color
  • Includes gromets