Learn More
MSC Justrite Plunger Can, 4 Quart, Steel, 1 ea
Catalog No. 5000675407
Encompass
Justrite plunger cans are designed to dispense flammable liquids and solvents safely, without waste or exposure.
  • Made of chemically resistant Ryton® and brass.
  • Pump base designed to leave less liquid in the bottom before refilling.
  • Pushing down on the pump assembly fills the upper pan with liquid.
  • Surplus liquid drains safely back into the reservoir.
  • Perforated upper pan serves as a flame arrester.
  • Top rim has a molded edge protector for smooth, trouble-free moistening.
Catalog No. 50-006-75407 Supplier MSC Supplier No. 89848204
Justrite plunger cans are designed to dispense flammable liquids and solvents safely, without waste or exposure.
  • Made of chemically resistant Ryton® and brass.
  • Pump base designed to leave less liquid in the bottom before refilling.
  • Pushing down on the pump assembly fills the upper pan with liquid.
  • Surplus liquid drains safely back into the reservoir.
  • Perforated upper pan serves as a flame arrester.
  • Top rim has a molded edge protector for smooth, trouble-free moistening.