Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Polypropylene Tray for 30- & 90-Minute 30-Gallon EN Cabinets

Catalog No. 19414057 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
30 gal. EN cabinets
45 gal. EN cabinets
Color:
Black
White
Product Type:
51mm curved edge
89mm curved edge
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Color Product Type
19-414-057 45 gal. EN cabinets Black 51mm curved edge
19-414-056 30 gal. EN cabinets White 89mm curved edge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-414-057 Supplier Justrite™ Supplier No. 22633
  • $423.00
    $285.65 / Each of 1

    Save $137.35

    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Extra layer of protection by utilizing polypropylene trays above steel shelves

  • Curved edge provides additional corrosion protection against leaks and spills
  • Fits 30- or 45-gal. EN cabinets

Specifications

Material Polypropylene
For Use With (Equipment) 45 gal. EN cabinets
Product Type 51mm curved edge
Color Black
Description Extra layer of protection by utilizing polypropylene trays above steel shelves
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.