Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Pop-Up Pools

Catalog No. 19111013 Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (English):
20 gal.
66 gal.
100 gal.
150 gal.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-111-013 150 gal.
19-111-010 20 gal.
19-111-011 66 gal.
19-111-012 100 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-111-013 Supplier Justrite™ Supplier No. 28325
Add to Cart
Add to Cart

To contain spills from punctured saddle tanks, leaking drums, hydraulic lines, pipes and machinery

  • Made of vinyl fabric
  • Color coded for visibility and easy identification
  • Folds tightly and stows easily
  • Embedded foam ring top raises containment wall as fluid level increases

Compliance

EPA for containment and spill prevention (SPCC).

Specifications

Color Yellow
Capacity (English) 150 gal.
Capacity (Metric) 568 L
Material PVC-coated Fabric
Type Pop-Up Pools
Height (English) 11 in.
Height (Metric) 27.9 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.