Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Portable Mixing Tank

Catalog No. 19037060 Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (English):
5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-037060 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-037060 Supplier Justrite™ Supplier No. 27705
  • $525.00
    $364.65 / Each of 1

    Save $160.35

    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal container for mixing operations, with fully removable cover

Easy to carry tank with swinging bail handle

  • Mixing tank minimizes the risk of spills or fire when mixing hazardous, flammable liquids
  • Self-closing, fully removable lid shuts securely and automatically ventilates the interior
  • Gasket seals tightly to prevent flammable vapors from escaping
  • Lid can be padlocked for added security
  • Internal flame arrester protects contents from ignition sources
  • Rugged 24-gauge steel construction with tough powder-coat finish provides maximum durability and chemical resistance
  • Use safety bonding tab to reduce static electricity buildup

Compliance

EPA, OSHA

Specifications

Certifications/Compliance EPA, OSHA
Product Type Portable Mixing Tank
Closure Type Removable Cap
Capacity (English) 5 gal.
Capacity (Metric) 18.9 L
Color Red
For Use With (Application) For Flammable Liquids
Material Steel
Finish Powder Coat
Includes Swinging bail handle, flame arrester, bonding tab
Height (English) 16 in.
Height (Metric) 406 mm
Diameter (English) Inner 15 in.
Diameter (Metric) Inner 381 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.