Learn More
Justrite™ Rolling Cart for Safety Cabinets
Catalog No. 19412629 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
Safety Cabinets
Catalog No. For Use With (Equipment)
19-412-629 Safety Cabinets
1 options
Catalog No. 19-412-629 Supplier Justrite™ Supplier No. 84001
  • $1,145.95
    $823.65 / Each of 1
    Save $322.30
    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Converts cabinet into mobile workstation to move materials to point-of-use
  • Fits any cabinet footprint of 43W x 18 in.D (109 x 45.7cm) with a maximum height of 44 in. (111.8cm), such as Justrite 30 gal. (114L) or piggyback cabinets
  • Two swivel and two fixed casters, each with brakes
  • Built-in grounding wire can be attached to cabinet
  • Ergonomic, removable push/pull handle
  • Black powder paint for durability
Specifications
Includes 2 Swivel and 2 fixed casters each with brakes, removable push or pull handle
For Use With (Equipment) Safety Cabinets
Height (English) 44 in.
Product Type Rolling Cart
For Use With (Application) To move materials to point-of-use
Height (Metric) 111.76 cm