Learn More
MSC Justrite Steel Head, 30 gal., FM UL, 1 ea
Catalog No. 5000789593
Encompass
This steel head is designed for Cease-Fire® waste receptacles, effectively smothering flames by cutting off the air supply. It provides continuous protection against trash fires for collecting waste paper and other solvent-free combustibles in various industrial and commercial settings.
  • Durable steel construction with red powder-coat paint
  • Unique contoured shape smothers flames fast
  • Ideal for abusive conditions
  • Suitable for 30-gallon drums
  • FM and UL approved
Catalog No. 50-007-89593 Supplier MSC Supplier No. 89848527
This steel head is designed for Cease-Fire® waste receptacles, effectively smothering flames by cutting off the air supply. It provides continuous protection against trash fires for collecting waste paper and other solvent-free combustibles in various industrial and commercial settings.
  • Durable steel construction with red powder-coat paint
  • Unique contoured shape smothers flames fast
  • Ideal for abusive conditions
  • Suitable for 30-gallon drums
  • FM and UL approved