Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Sure-Grip™ EX™ Safety Cabinets With Handles, For Flammables
SCI_ED

Catalog No. S04760MF Shop All Justrite Safety Group Products
Change view
Click to view available options
Load Bearing Capacity:
30 gal.
45 gal.
Door Style:
Manual-close
Self-close
No. of Doors:
1
2
No. of Shelves:
1
2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Load Bearing Capacity Door Style No. of Doors No. of Shelves
S04760MF 30 gal. Manual-close 2 1
S04762MF 45 gal. Self-close 1 2
S04761MF 45 gal. Manual-close 2 2
S04763MF 45 gal. Self-close 2 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S04760MF Supplier Justrite™ Supplier No. 893000/EMD

Nonslip gripper allows fingertip opening.

  • Sure-Grip EX flush-mounted door handle with two keys for security
  • Handle can be padlocked (padlock not included), no interference with traffic
  • Exclusive self-latching feature ensures safe closing of cabinet doors without manually engaging door handle
  • Double-wall construction provides 1.5 in. (3.8cm) of insulating air space to help prevent spread of flames
  • High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
  • Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self-latch engages critical three-point latching system for protection under fire conditions
  • Two-door models hinged left and right with self-closing mechanism ensures proper closing sequence
  • Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 165°F (74°C )
  • Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
  • SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Adjustable leveling feet ensure solid positioning and alignment for tight door closure
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • Meet OSHA and NFPA requirements
  • Color: Yellow

Certifications

FM-approved

Compliance

Meet NFPA Code 30 and OSHA regulations; models with self-close doors also comply with the International Fire Code and NFPA1, FIRE CODE™.

Warranty and Services

10 years

Specifications

Dimensions (D x W x H) Interior 14.56 x 39.5 x 39.62 in.
Product Type Flammables Safety Cabinet
Height (Metric) Exterior 112 cm
Width (Metric) Exterior 109 cm
Depth (Metric) Interior 37 cm
No. of Shelves 1
Depth (Metric) Exterior 45.7 cm
No. of Doors 2
Height (Metric) Interior 100.6 cm
Width (Metric) Interior 100.3 cm
Weight (Metric) 105 kg
Load Bearing Capacity 30 gal.
Surface Coating Epoxy, Polyester
Depth (English) Exterior 18 in.
Depth (English) Interior 14.56 in.
Door Style Manual-close
Certifications/Compliance FM, NFPA, OSHA
Height (English) Exterior 44 in.
Width (English) Interior 39.5 in.
Height (English) Interior 39.62 in.
Includes U-Loc™ padlockable handle, Haz-Alert™ reflective trilingual warning labels, sturdy SpillSlope™ shelves
Weight (English) 231 lb.
Color Yellow
For Use With (Application) Flammable liquid storage and protection
Material Steel (CR)
Width (English) Exterior 43 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.