Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Sure-Grip™ EX Slimline Hazardous Material Safety Cabinet
GREENER_CHOICE

Catalog No. p-4817171 Shop All Justrite Safety Group Products
Change view
Click to view available options
Load Bearing Capacity:
22 gal.
54 gal.
Dimensions (D x W x H):
18 x 23.25 x 65 in.
34 x 23.25 x 65 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Load Bearing Capacity Dimensions (D x W x H)
19-414-807 22 gal. 18 x 23.25 x 65 in.
19-414-808 54 gal. 34 x 23.25 x 65 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-414-807 Supplier Justrite™ Supplier No. 862228
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Keep storage of your hazardous materials compliant with the Justrite™ Sure-Grip™ EX Hazmat Safety Cabinet.

  • Double-wall construction, dual vents, grounding wire connections, adjustable shelves
  • Leakproof sills, three-point self-latching doors and leveling feet
  • Self-close door(s) shuts and latches automatically when fusible link melts at 74°C (165°F) under fire conditions
  • Unique, concealed self-closing mechanism offers obstruction-free access to contents
  • Include polyethylene trays that sit on top of galvanized steel shelves and separate polyethylene liner for bottom sump to resist aggressive chemicals
  • Easily remove liner for quick cleaning of drips and leaks
  • Epoxy baked-on powder-coat finish, inside and out, provides increased chemical resistance

Specifications

Product Type Combi Safety Storage Cabinet
Dimensions (D x W x H) 18 x 23.25 x 65 in.
Door Style Self-close
No. of Doors 1
No. of Shelves 3
Load Bearing Capacity 22 gal.
Color Blue
For Use With (Application) Hazardous materials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.