Learn More
Cayman Chemical JuvenIle Hormon III 10mg
SDP
Catalog No. 502982833
Encompass_Preferred
An acyclic sesquiterpenoid that regulates diverse processes in insects; activates the juvenile hormone receptor, known as methoprene-tolerant, with a Kd value of 2.9 nM
Catalog No. 50-298-2833 Supplier Cayman Chemical Supplier No. 1964610 MG
An acyclic sesquiterpenoid that regulates diverse processes in insects; activates the juvenile hormone receptor, known as methoprene-tolerant, with a Kd value of 2.9 nM