Learn More
Medchemexpress LLC JV-1-36 acetate | 221377-79-3 | 99.93% | 3785.88 (free base) | 5 MG
SDP
Catalog No. 5000399290
Encompass_Preferred
JV-1-36 acetate is a growth hormone-releasing hormone (GHRH) antagonist that demonstrates antioxidant effects, inhibiting reactive oxygen species in A549 lung cancer cells and reducing cell viability in A549 and Hela cells. It is suitable for in vitro research on GHRH antagonists.
  • GHRH antagonist
  • Inhibits reactive oxygen species in A549 lung cancer cells
  • Reduces cell viability in A549 and Hela cells
  • Suitable for in vitro research on GHRH antagonists
  • Exerts antioxidant effects
Catalog No. 50-003-99290 Supplier Medchemexpress LLC Supplier No. HYP3397A5MG
JV-1-36 acetate is a growth hormone-releasing hormone (GHRH) antagonist that demonstrates antioxidant effects, inhibiting reactive oxygen species in A549 lung cancer cells and reducing cell viability in A549 and Hela cells. It is suitable for in vitro research on GHRH antagonists.
  • GHRH antagonist
  • Inhibits reactive oxygen species in A549 lung cancer cells
  • Reduces cell viability in A549 and Hela cells
  • Suitable for in vitro research on GHRH antagonists
  • Exerts antioxidant effects