Learn More
Medchemexpress LLC HY-13968 10mg Medchemexpress, JW 55 CAS:664993-53-7 Purity:>98%
SDP
Catalog No. 502081538
Encompass_Preferred
Medchemexpress, HY-13968 10mg JW 55 CAS:664993-53-7 JW 55 is a potent and selective β-catenin signaling pathway inhibitor, which functions via inhibition of the PARP domain of tankyrase 1 and tankyrase 2 (TNKS1/2). JW 55 decreases auto-PARsylation of TNKS1/2 in vitro with IC50s of 1.9 μM and 830 nM respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1538 Supplier Medchemexpress LLC Supplier No. HY1396810MG
Medchemexpress, HY-13968 10mg JW 55 CAS:664993-53-7 JW 55 is a potent and selective β-catenin signaling pathway inhibitor, which functions via inhibition of the PARP domain of tankyrase 1 and tankyrase 2 (TNKS1/2). JW 55 decreases auto-PARsylation of TNKS1/2 in vitro with IC50s of 1.9 μM and 830 nM respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.