Learn More
Apexbio Technology LLC JW 55 664993-53-7 10mg
SDP
Catalog No. 502240960
Encompass_Preferred
JW 55 (CAS 664993-53-7) is a small-molecule inhibitor that targets the poly(ADP-ribose) polymerase (PARP) domains of tankyrase 1 and tankyrase 2 (TNKS1/2) with reported IC50 values of 1 9 M and 0 83 M respectively By inhibiting TNKS1/2 activity JW 55 stabilizes AXIN2 a component of the -catenin destruction complex leading to enhanced degradation of -catenin and suppression of canonical Wnt/ -catenin signaling In vitro JW 55 suppresses Wnt pathway activity in colorectal cancer cells harboring APC or -catenin mutations In vivo it reduces Wnt-driven phenotypes in Xenopus embryos and diminishes polyp formation in APC-mutant mice supporting its utility as a research tool in Wnt/ -catenin signaling and cancer biology
This item is not returnable. View return policy
Catalog No. 50-224-0960 Supplier Apexbio Technology LLC Supplier No. A452910
JW 55 (CAS 664993-53-7) is a small-molecule inhibitor that targets the poly(ADP-ribose) polymerase (PARP) domains of tankyrase 1 and tankyrase 2 (TNKS1/2) with reported IC50 values of 1 9 M and 0 83 M respectively By inhibiting TNKS1/2 activity JW 55 stabilizes AXIN2 a component of the -catenin destruction complex leading to enhanced degradation of -catenin and suppression of canonical Wnt/ -catenin signaling In vitro JW 55 suppresses Wnt pathway activity in colorectal cancer cells harboring APC or -catenin mutations In vivo it reduces Wnt-driven phenotypes in Xenopus embryos and diminishes polyp formation in APC-mutant mice supporting its utility as a research tool in Wnt/ -catenin signaling and cancer biology