Learn More
TARGETMOL CHEMICALS INC JW 55 25MG
SDP
Catalog No. 502577296
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JW 55 (JW55) is an effective and selective (beta)-catenin signaling pathway inhibitor works by inhibition of the PARP domain of tankyrase 1 and tankyrase 2 (TNKS1/2). purity: 99%
Catalog No. 50-257-7296 Supplier TARGETMOL CHEMICALS INC Supplier No. T180725MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JW 55 (JW55) is an effective and selective (beta)-catenin signaling pathway inhibitor works by inhibition of the PARP domain of tankyrase 1 and tankyrase 2 (TNKS1/2). purity: 99%