Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JW 642, Tocris Bioscience™

Catalog No. 490610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-061-0 10 mg
49-065-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 49-061-0 Supplier R&D Systems Supplier No. 4906/10
This item is not returnable. View return policy

MAGL inhibitor

Potent and selective monoacylglycerol lipase (MAGL) inhibitor (IC50 = 3.7 nM). Displays >1000-fold selectivity for MAGL over fatty acid amide hydrolase (IC50 = 20.6 μM). Analog of JZL 195 (Cat. No. 4715).

Chemical Identifiers

CAS 1416133-89-5
Molecular Formula C21H20F6N2O3
Molecular Weight (g/mol) 462.392
InChI Key AVSCNEOUWSVZEY-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoropropan-2-yl 4-3-phenoxybenzyl piperazine-1-carboxylate, 1,1,1,3,3,3-hexafluoropropan-2-yl 4-3-phenoxyphenyl methyl piperazine-1-carboxylate, 4-3-phenoxyphenyl methyl-1-piperazinecarboxylic acid 2,2,2-trifluoro-1-trifluoromethyl ethyl ester
PubChem CID 71656520
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-yl 4-[(3-phenoxyphenyl)methyl]piperazine-1-carboxylate
SMILES C1CN(CCN1CC2=CC(=CC=C2)OC3=CC=CC=C3)C(=O)OC(C(F)(F)F)C(F)(F)F

Specifications

Color White
Quantity 10 mg
Formula Weight 462.39
Percent Purity >98%
Physical Form Solid
Chemical Name or Material JW 642
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.