Learn More
Medchemexpress LLC JW67 | 442644-28-2 | 98.15% | 394.38 | 50 MG
SDP
Catalog No. 5000346756
Encompass_Preferred
JW67 is a Wnt Signaling Inhibitor with an IC50 of 1.17 μM. It impacts the multiprotein complex of β-catenin/GSK-3β/AXIN/APC/CK1, leading to a rapid reduction in active β-catenin and downregulation of Wnt target genes. It also inhibits colorectal cancer cell growth.
  • Inhibits canonical Wnt signaling
  • Reduces active β-catenin
  • Downregulates Wnt target genes
  • Inhibits colorectal cancer cell growth
  • Increases AXIN2 concentration
  • Affects Wnt target gene expression
  • Reduces cell growth in colon cancer cell lines
Catalog No. 50-003-46756 Supplier Medchemexpress LLC Supplier No. HY10844210MG
JW67 is a Wnt Signaling Inhibitor with an IC50 of 1.17 μM. It impacts the multiprotein complex of β-catenin/GSK-3β/AXIN/APC/CK1, leading to a rapid reduction in active β-catenin and downregulation of Wnt target genes. It also inhibits colorectal cancer cell growth.
  • Inhibits canonical Wnt signaling
  • Reduces active β-catenin
  • Downregulates Wnt target genes
  • Inhibits colorectal cancer cell growth
  • Increases AXIN2 concentration
  • Affects Wnt target gene expression
  • Reduces cell growth in colon cancer cell lines