Learn More
Medchemexpress LLC JWG-071 | 2250323-50-1 | 99.15% | 612.76 | 10 MG
SDP
Catalog No. 5000344688
Encompass_Preferred
JWG-071 is a kinase-selective chemical probe for ERK5. It inhibits ERK5 and LRRK2 with IC50 values of 88 nM and 109 nM, respectively. This compound is useful for research in the MAPK/ERK pathway and Stem Cell/Wnt pathways.
  • Kinase-selective chemical probe for ERK5.
  • Inhibits ERK5 with an IC50 of 88 nM.
  • Inhibits LRRK2 with an IC50 of 109 nM.
  • Available in various solid quantities.
  • High purity.
  • Can be dissolved in DMSO for in vitro studies.
  • Suitable for in vivo research with specific formulation recommendations.
Catalog No. 50-003-44688 Supplier Medchemexpress LLC Supplier No. HY108886100MG
JWG-071 is a kinase-selective chemical probe for ERK5. It inhibits ERK5 and LRRK2 with IC50 values of 88 nM and 109 nM, respectively. This compound is useful for research in the MAPK/ERK pathway and Stem Cell/Wnt pathways.
  • Kinase-selective chemical probe for ERK5.
  • Inhibits ERK5 with an IC50 of 88 nM.
  • Inhibits LRRK2 with an IC50 of 109 nM.
  • Available in various solid quantities.
  • High purity.
  • Can be dissolved in DMSO for in vitro studies.
  • Suitable for in vivo research with specific formulation recommendations.