Learn More
Medchemexpress LLC JWG-071 | 2250323-50-1 | 99.2% | 612.76 | 10 MG
SDP
Catalog No. 5000344924
Encompass_Preferred
JWG-071 is a kinase-selective chemical probe for Extracellular signal-regulated kinase 5 (ERK5). This compound demonstrates potent inhibitory activity against key kinases, making it a valuable tool for research applications.
  • Inhibits ERK5 with an IC50 value of 88 nM.
  • Inhibits Leucine-rich repeat kinase 2 (LRRK2) with an IC50 value of 109 nM.
Catalog No. 50-003-44924 Supplier Medchemexpress LLC Supplier No. HY10888650MG
JWG-071 is a kinase-selective chemical probe for Extracellular signal-regulated kinase 5 (ERK5). This compound demonstrates potent inhibitory activity against key kinases, making it a valuable tool for research applications.
  • Inhibits ERK5 with an IC50 value of 88 nM.
  • Inhibits Leucine-rich repeat kinase 2 (LRRK2) with an IC50 value of 109 nM.