Learn More
Medchemexpress LLC HY-108886 5mg Medchemexpress, JWG-071 CAS:2250323-50-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1088865MG
Encompass_Preferred
Medchemexpress, HY-108886 5mg JWG-071 CAS:2250323-50-1 JWG-071 is the first reported kinase-selective chemical probe for ERK5. JWG-071 improves ERK5 activity and BRD4 selectivity. JWG-071 will be a much-needed chemical probe for deconvoluting ERK5 and BRD4 pharmacology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8631