Learn More
Cayman Chemical JWH 007d9 GC MS Standards
SDP
Catalog No. 502889690
Encompass_Preferred
An internal standard for the quantification of JWH 007 by GC- or LC-MS; intended for forensic and research purposes
Catalog No. 50-288-9690 Supplier Cayman Chemical Supplier No. 10486500 UG
An internal standard for the quantification of JWH 007 by GC- or LC-MS; intended for forensic and research purposes