Learn More
Cayman Chemical JWH 007d9 GC MS Standards
SDP
Catalog No. 502888376
Encompass_Preferred
An internal standard for the quantification of JWH 007 by GC- or LC-MS; intended for forensic and research purposes
Catalog No. 50-288-8376 Supplier Cayman Chemical Supplier No. 104861 MG
Add to Cart
Add to Cart
An internal standard for the quantification of JWH 007 by GC- or LC-MS; intended for forensic and research purposes