Learn More
Cayman Chemical JWH 015d7 GC MS Standards
SDP
Supplier:  Cayman Chemical 106601 MG
Encompass_Preferred
An internal standard for the quantification of JWN 015 by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-289-0238