Learn More
Cayman Chemical JWH 015d7 GC MS Standards
SDP
Catalog No. 502889668
Encompass_Preferred
An internal standard for the quantification of JWN 015 by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-288-9668 Supplier Cayman Chemical Supplier No. 10660500 UG
An internal standard for the quantification of JWN 015 by GC- or LC-MS; intended for research and forensic applications