Learn More
Cayman Chemical JWH 018 2'-naphthyl isomer, Purity: ≥98%, CAS: 1131605-25-8, 5 mg
SDP
Catalog No. 502892710
Encompass_Preferred
Differs structurally from JWH 018 by having the naphthyl group attached at the 2' position
Catalog No. 50-289-2710 Supplier Cayman Chemical Supplier No. 90010045 MG
Differs structurally from JWH 018 by having the naphthyl group attached at the 2' position