Learn More
Cayman Chemical JWH 018 2'-naphthyl-N-(1,2-dimethylpropyl) isomer, Purity: ≥98%, CAS: 1870862-08-0, 10 mg
SDP
Catalog No. 502895725
Encompass_Preferred
Differs from JWH 018 structurally by having a 1,2-dimethylpropyl chain, rather than a pentyl chain, extending from the indole group and the naphthyl moiety attached at its 2 position
Catalog No. 50-289-5725 Supplier Cayman Chemical Supplier No. 900100610 MG
Differs from JWH 018 structurally by having a 1,2-dimethylpropyl chain, rather than a pentyl chain, extending from the indole group and the naphthyl moiety attached at its 2 position