Learn More
Cayman Chemical JWH 018 2naphthylN 1ethy 10mg
SDP
Catalog No. 502951772
Encompass_Preferred
An isomer of JWH 018, differing by having the naphthyl group attached at the 2, rather than 1, position, as well as an ethylpropyl group in place of the pentyl chain; intended for research and forensic
Catalog No. 50-295-1772 Supplier Cayman Chemical Supplier No. 1158610 MG
An isomer of JWH 018, differing by having the naphthyl group attached at the 2, rather than 1, position, as well as an ethylpropyl group in place of the pentyl chain; intended for research and forensic