Learn More
Cayman Chemical JWH 018 6-methoxyindole analog, Purity: ≥98%, CAS: 1427325-49-2, 5 mg
SDP
Catalog No. 502888277
Encompass_Preferred
Analog of JWH 018 6-hydroxyindole metabolite, a urinary metabolite of the CB receptor agonist JWH 018
Catalog No. 50-288-8277 Supplier Cayman Chemical Supplier No. 106975 MG
Analog of JWH 018 6-hydroxyindole metabolite, a urinary metabolite of the CB receptor agonist JWH 018