Learn More
Cayman Chemical JWH 018 8quInolnyl caram 10mg
SDP
Supplier:  Cayman Chemical 1489110 MG
Encompass_Preferred
A structural analog of JWH 018 in which the naphthalenyl-methanone portion has been replaced by 8-quinolinyl carboxamide; intended for research and forensic applications
Catalog No. 50-296-9514