Learn More
Cayman Chemical JWH 018 benzimidazole analog
SDP
Catalog No. 502971111
Encompass_Preferred
Replaces the indole core of JWH 018 with a benzimidazole core, which may confer higher selectivity for the CB2 receptor; intended for forensic and research applications
Catalog No. 50-297-1111 Supplier Cayman Chemical Supplier No. 150745 MG
Replaces the indole core of JWH 018 with a benzimidazole core, which may confer higher selectivity for the CB2 receptor; intended for forensic and research applications