Learn More
Cayman Chemical JWH 018 benzimidazole analog
SDP
Supplier:  Cayman Chemical 150741 MG
Encompass_Preferred
Replaces the indole core of JWH 018 with a benzimidazole core, which may confer higher selectivity for the CB2 receptor; intended for forensic and research applications
Catalog No. 50-297-0901