Learn More
Cayman Chemical JWH 018 N 1 2dImthylPRYl 10mg
SDP
Catalog No. 502892593
Encompass_Preferred
Differs from JWH 018 structurally by having a 1,2-dimethylpropyl chain, rather than a pentyl chain, extending from the indole group; intended for forensic purposes
Catalog No. 50-289-2593 Supplier Cayman Chemical Supplier No. 900100310 MG
Differs from JWH 018 structurally by having a 1,2-dimethylpropyl chain, rather than a pentyl chain, extending from the indole group; intended for forensic purposes