Learn More
Cayman Chemical JWH 018 N-(2,2-dimethylpropyl) isomer, Purity: ≥98%, CAS: 1427325-47-0, 10 mg
SDP
Catalog No. 502894129
Encompass_Preferred
Differs from JWH 018 structurally by having a 2,2-dimethylpropyl group, rather than a pentyl chain, extending from the indole group; intended for research and forensic applications
Catalog No. 50-289-4129 Supplier Cayman Chemical Supplier No. 900100110 MG
Differs from JWH 018 structurally by having a 2,2-dimethylpropyl group, rather than a pentyl chain, extending from the indole group; intended for research and forensic applications