Learn More
Cayman Chemical JWH 018 N 3methylbutyl isomer
SDP
Catalog No. 502891422
Encompass_Preferred
An analog of JWH 018, a selective agonist of the CB1 receptor; intended for research and forensic applications
Catalog No. 50-289-1422 Supplier Cayman Chemical Supplier No. 1069110 MG
An analog of JWH 018, a selective agonist of the CB1 receptor; intended for research and forensic applications