Learn More
Cayman Chemical JWH 018 N 4hydroxypenty 100ug
SDP
Catalog No. 502951514
Encompass_Preferred
An internal standard for the quantifiction of (±)-JWH 018 N-(4-hydroxypentyl) metabolite by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-295-1514 Supplier Cayman Chemical Supplier No. 10926100 UG
An internal standard for the quantifiction of (±)-JWH 018 N-(4-hydroxypentyl) metabolite by GC- or LC-MS; intended for research and forensic applications