Learn More
Cayman Chemical JWH 018 N-(5-hydroxypentyl) metabolite, Purity: ≥98%, CAS: 335161-21-2, 10 mg
SDP
Catalog No. 502895598
Encompass_Preferred
An analytical reference standard categorized as a synthetic cannabinoid; an active metabolite of JWH 018 and AM2201; intended for research and forensic applications
Catalog No. 50-289-5598 Supplier Cayman Chemical Supplier No. 900085510 MG
An analytical reference standard categorized as a synthetic cannabinoid; an active metabolite of JWH 018 and AM2201; intended for research and forensic applications