Learn More
Cayman Chemical JWH 018 N 5bromopentyl na 5mg
SDP
Supplier:  Cayman Chemical 110475 MG
Encompass_Preferred
An analytical reference standard that is structurally similar to known synthetic cannabinoids; intended for research and forensic applications
Catalog No. 50-295-1388