Learn More
Cayman Chemical JWH 018 N-(5-chloropentyl) analog, Purity: ≥95%, CAS: 1445578-56-2, 1 mg
SDP
Catalog No. 502888254
Encompass_Preferred
Differs structurally from JWH 018 by having chlorine added to the 5 position of the pentyl chain; intended for forensic and research applications
Catalog No. 50-288-8254 Supplier Cayman Chemical Supplier No. 105211 MG
Differs structurally from JWH 018 by having chlorine added to the 5 position of the pentyl chain; intended for forensic and research applications