Learn More
Cayman Chemical JWH 018 N-pentanoic acid metabolite, Purity: ≥95%, CAS: 1254475-87-0, 10 mg
SDP
Catalog No. 502895209
Encompass_Preferred
An analytical reference standard categorized as a synthetic cannabinoid metabolite; a metabolite of JWH 018 and AM2201; intended for research and forensic applications
Catalog No. 50-289-5209 Supplier Cayman Chemical Supplier No. 900085610 MG
An analytical reference standard categorized as a synthetic cannabinoid metabolite; a metabolite of JWH 018 and AM2201; intended for research and forensic applications