Learn More
Cayman Chemical JWH 018 NpentnoIc ACD meta
SDP
Catalog No. 502893004
Encompass_Preferred
An internal standard for the quantification of JWH 018 N-pentanoic acid metabolite by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-289-3004 Supplier Cayman Chemical Supplier No. 9000867500 UG
An internal standard for the quantification of JWH 018 N-pentanoic acid metabolite by GC- or LC-MS; intended for research and forensic applications