Learn More
Cayman Chemical JWH 019 5-hydroxyindole metabolite, Purity: ≥98%, CAS: 1379604-70-2, 10 mg
SDP
Catalog No. 502893173
Encompass_Preferred
Expected to be a major metabolite, detectable in serum and urine, of JWH 019, based on the metabolism of the closely related compounds JWH 015 and JWH 018; biological effects are not known; intended for research and forensic applications
Catalog No. 50-289-3173 Supplier Cayman Chemical Supplier No. 900076410 MG
Expected to be a major metabolite, detectable in serum and urine, of JWH 019, based on the metabolism of the closely related compounds JWH 015 and JWH 018; biological effects are not known; intended for research and forensic applications