Learn More
Cayman Chemical JWH 019 N 5hydroxyhexyl me
SDP
Catalog No. 502971460
Encompass_Preferred
An internal standard for the quantification of JWH 019 N-(5-hydroxyhexyl) metabolite by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-297-1460 Supplier Cayman Chemical Supplier No. 14687100 UG
An internal standard for the quantification of JWH 019 N-(5-hydroxyhexyl) metabolite by GC- or LC-MS; intended for research and forensic applications