Learn More
Cayman Chemical JWH 019 N 5hydroxyhexyl me
SDP
Supplier:  Cayman Chemical 14687500 UG
Encompass_Preferred
An internal standard for the quantification of JWH 019 N-(5-hydroxyhexyl) metabolite by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-297-1935