Learn More
Cayman Chemical JWH 019 N 5hydroxyhexyl me 5mg
SDP
Catalog No. 502972396
Encompass_Preferred
A urinary metabolite of JWH 019, detectable both in serum and urine; intended for forensic and research applications
Catalog No. 50-297-2396 Supplier Cayman Chemical Supplier No. 148145 MG
A urinary metabolite of JWH 019, detectable both in serum and urine; intended for forensic and research applications