Learn More
Cayman Chemical JWH 019 N-(6-hydroxyhexyl) metabolite, Purity: ≥98%, CAS: 1435934-29-4, 10 mg
SDP
Catalog No. 502895878
Encompass_Preferred
An expected metabolite of JWH 019, detectable both in serum and urine; intended for research and forensic applications
Catalog No. 50-289-5878 Supplier Cayman Chemical Supplier No. 900076510 MG
An expected metabolite of JWH 019, detectable both in serum and urine; intended for research and forensic applications