Learn More
Cayman Chemical JWH 031 Analytical Reference S
SDP
Supplier:  Cayman Chemical 1082410 MG
Encompass_Preferred
A synthetic CB with a relatively low binding affinity for the central CB1 receptor (Ki = 399 nM); efficacious in reducing spontaneous activity and increasing antinociception in mice; intended for forensic and research applications
Catalog No. 50-289-1377