Learn More
Cayman Chemical JWH 031 Analytical Reference S
SDP
Catalog No. 502887812
Encompass_Preferred
A synthetic CB with a relatively low binding affinity for the central CB1 receptor (Ki = 399 nM); efficacious in reducing spontaneous activity and increasing antinociception in mice; intended for forensic and research applications
Catalog No. 50-288-7812 Supplier Cayman Chemical Supplier No. 108245 MG
A synthetic CB with a relatively low binding affinity for the central CB1 receptor (Ki = 399 nM); efficacious in reducing spontaneous activity and increasing antinociception in mice; intended for forensic and research applications