Learn More
Cayman Chemical JWH 073 2'-naphthyl isomer, Purity: ≥97%, CAS: 2365471-75-4, 1 mg
SDP
Catalog No. 502893616
Encompass_Preferred
Differs structurally from JWH 073 by having the naphthyl group attached at the 2' position
Catalog No. 50-289-3616 Supplier Cayman Chemical Supplier No. 90010141 MG
Differs structurally from JWH 073 by having the naphthyl group attached at the 2' position