Learn More
Cayman Chemical JWH 073 2naphthylN 2meth 10mg
SDP
Catalog No. 502895726
Encompass_Preferred
Differs structurally from JWH 073 by having the naphthyl group attached at the 2' position and a 2-methylpropyl group in place of a butyl chain
Catalog No. 50-289-5726 Supplier Cayman Chemical Supplier No. 900101510 MG
Differs structurally from JWH 073 by having the naphthyl group attached at the 2' position and a 2-methylpropyl group in place of a butyl chain