Learn More
Cayman Chemical JWH 073 4-methylnaphthyl anal
SDP
Supplier:  Cayman Chemical 90010765 MG
Encompass_Preferred
Differs structurally from JWH 073 by having a methyl group added at the 4 position of the naphthyl rings; a similar addition to JWH 018 results in an analog which has higher affinities for both cannabinoid receptors
Catalog No. 50-289-5691