Learn More
Cayman Chemical JWH 073 4-methylnaphthyl analog, Purity: ≥98%, CAS: 1354631-21-2, 1 mg
SDP
Catalog No. 502895690
Encompass_Preferred
Differs structurally from JWH 073 by having a methyl group added at the 4 position of the naphthyl rings; a similar addition to JWH 018 results in an analog which has higher affinities for both cannabinoid receptors
Catalog No. 50-289-5690 Supplier Cayman Chemical Supplier No. 90010761 MG
Differs structurally from JWH 073 by having a methyl group added at the 4 position of the naphthyl rings; a similar addition to JWH 018 results in an analog which has higher affinities for both cannabinoid receptors